Sorvall™ Legend™ X1 Centrifuge Series
Sorvall™ Legend™ X1 Centrifuge Series
Actual product may vary
Sorvall™ Legend™ X1 Centrifuge Series
Sorvall™ Legend™ X1 Centrifuge Series
Sorvall™ Legend™ X1 Centrifuge Series
Sorvall™ Legend™ X1 Centrifuge Series
Sorvall™ Legend™ X1 Centrifuge Series
Process diverse laboratory applications with the high-performance Thermo Scientific™ Sorvall™ Legend™ X1 Centrifuge Series.
Have Questions?
Catalog NumberRefrigeratedElectrical RequirementsDescription
75004220No230 V 50/60 HzSorvall Legend X1 Centrifuge
Catalog number 75004220
Price (SGD)
-
Refrigerated:
No
Electrical Requirements:
230 V 50/60 Hz
Description:
Sorvall Legend X1 Centrifuge
Process diverse laboratory applications with the high-performance Thermo Scientific™ Sorvall™ Legend™ X1 Centrifuge Series, which offers a broad range of rotors and adapters, including lightweight Thermo Scientific™ Fiberlite™ carbon fiber rotors.
Simple Operation
  • Intuitive control panel and digital display
  • Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection
  • Easily adjust speed, time and temperature, even during the run with our convenient keyboard for direct input
  • Innovative lid-locking technology enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches with optional automatic lid opening
  • Double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run
Auto-Lock™ III Rotor System
  • Secure locking system allows easy push-button installation and exchange of rotors
  • Saves time when switching rotors between applications
  • Easy access to the chamber enables quick cleaning, a healthier working environment and longer unit life
ClickSeal™ Bucket Sealing System
  • Biocontainment system seals with a snap; eliminates screw caps and complicated clips
  • Glove-friendly, one-handed open/close capability
  • Certified by CAMR™ in Porton Down, UK
SMARTSpin™ Technology
  • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option
  • Optimizes safety and improves separations
Space-Saving Design
  • Compact size conserves workspace
  • Ergonomic height and curved design ensure comfortable sample handling and cleaning of unit
Energy Efficient
  • Design saves up to 40% of energy on industry standard protocols such as blood separations or conical-tube processing
Ventilated Sorvall Legend X1
  • Dimensions: 23.8L × 17.3W × 14.2 in.H [34.3 in.H open] (60.5 × 44 × 36cm [87cm open])
  • Net weight: 126.8 lb. (57.5kg)
Refrigerated Sorvall Legend X1R
  • CFC-free refrigeration system
  • Temperature Range: -10° to +40°C
  • Pre-Cooling Function with direct button
  • 23.8L × 24.5W × 14.2 in.H [34.3 in.H open] (60.5 × 62.3 × 36cm [87cm open])
  • Net weight: 201.7 lb. (91.5kg)
Ordering Information:
For exceptional versatility, the Sorvall X1 series supports a wide range of rotors and adapters, including Thermo Scientific™ Fiberlite™ rotors, for a minimum of 10 rotor configurations. All rotors and adapters are sold separately.
Recommended for:
Cell culture, microplate, bioproduction, food safety, blood separation, bodily fluids processing, pharmaceutical, biotechnology, stem cell, microbiology, molecular biology, proteomics, genomics
Specifications
Certifications/ComplianceIVD compliant; IEC 61010-1, IEC 61010-2-020, IEC 61010-2-101; 230V models meet EN 292, EN 61326, EN 55011B. UL, CE, CSA, Certified Biosafety
Controller TypeMicroprocessor
DescriptionSorvall Legend X1 Centrifuge
Capacity4 x 400 mL
Max. RCF25,830 x g
Max. Speed15,200 rpm
Product LineSorvall
RefrigeratedNo
DisplayDigital
Drive SystemDirect, Brushless Induction Low Profile
Noise Level<61 dB(A)
Program Storage99 programs (5 with direct access key)
Run Time99 hr. 59 min. plus HOLD
TypeVentilated Centrifuge
Electrical Requirements230 V 50/60 Hz