Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Actual product may vary
Buckets for Thermo Scientific™ HAEMAFlex™ Swinging Bucket Rotors
Expand individual bucket capacity or maximize total blood bags per spin with choice of buckets for Thermo Scientific™ HAEMAFlex™ blood banking rotors.
Have Questions?
Catalog NumberFor Use With (Equipment)Description
75003846HAEMAFlex 12/16 Swing Bucket RotorDouble Bucket
Catalog number 75003846
HSN 84219100
Price (SGD)
4,983.00
Special offer
Ends: 30-Jun-2026
5,862.00
Save 879.00 (15%)
Each
For Use With (Equipment):
HAEMAFlex 12/16 Swing Bucket Rotor
Description:
Double Bucket
Price (SGD)
4,983.00
Special offer
Ends: 30-Jun-2026
5,862.00
Save 879.00 (15%)
Each
Thermo Scientific blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
For high-throughput blood processing needs, the HAEMAFlex 12 swinging bucket rotor provides 12 x 500mL capacity or upgrade your centrifuge capacity to 16 x 500mL blood bags with the HAEMAFlex 16 rotor and the same bucket selections.
Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Specifications
For Use With (Equipment)HAEMAFlex 12/16 Swing Bucket Rotor
Capacity16 x 500 mL
TypeDouble Blood Bag Bucket
DescriptionDouble Bucket
Unit SizeEach