Adapters for Single Blood Bag Bucket with Filter Pack for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Adapters for Single Blood Bag Bucket with Filter Pack for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Actual product may vary
Adapters for Single Blood Bag Bucket with Filter Pack for Thermo Scientific™ HAEMAFlex™ 6/8 Rotors
Accommodate 450–550mL Triple, Quad or Quint blood bags with additive solution in single blood bag buckets with filter pack for Thermo Scientific™ HAEMAFlex™6/8 rotors.
Have Questions?
Catalog NumberType
75003842Blood Bag Adapter
Catalog number 75003842
Price (SGD)
529.00
EA
Add to cart
Type:
Blood Bag Adapter
Price (SGD)
529.00
EA
Add to cart
Thermo Scientific™ blood banking centrifuges offer the flexibility to select higher capacity rotors or match existing workflows, with a choice of Thermo Scientific™ HAEMAFlex™ rotors and bucket systems.
Address your lab’s regular- to medium-throughput blood processing needs with the HAEMAFlex 6 swinging bucket rotor with 6 x 550mL blood bag capacity or, utilizing the same bucket system, upgrade to 8 x 550mL bags with the HAEMAFlex 8 swinging bucket rotor.
Select an adapter for 450–550mL Triple, Quad or Quint blood bag with additive solution.
Specifications
TypeBlood Bag Adapter
Capacity6/8 x 550 mL
For Use With (Equipment)Single blood bag bucket with filter pack
Description450-550mL Triple, Quad or Quint Blood Bag with Additive Solution Adapter (set of 2)
Unit SizeEA