CLINIConic™ Rotor Adapters
CLINIConic™ Rotor Adapters
Actual product may vary
CLINIConic™ Rotor Adapters
CLINIConic™ Rotor Adapters
CLINIConic™ Rotor Adapters
CLINIConic™ Rotor Adapters
CLINIConic™ Rotor Adapters
Incorporate these rotor adapters with the CLINIConic fixed-angle rotor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500370210 mL Round-bottom Tube
111725955 mL Round-bottom or Blood Collection Tube
111725967 mL Round-bottom or Blood Collection Tube
Catalog number 75003702
Price (SGD)
170.00
EA
Add to cart
Capacity:
10 mL Round-bottom Tube
Price (SGD)
170.00
EA
Add to cart
Incorporate Thermo Scientific™ CLINIConic™ Rotor Adapters with the CLINIConic fixed-angle rotor.
Ordering Information:
CLINIConic fixed-angle rotor body (75-003-623) is available separately. Contact your sales representative for more information.
Specifications
TypeRotor Tube Adapter
Capacity10 mL Round-bottom Tube
Unit SizeEA