Fiberlite F15-6 x 100y Fixed Angle Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Actual product may vary
Fiberlite F15-6 x 100y Fixed Angle Rotor for 1.6 L and 4 L General Purpose X Pro and ST Plus Centrifuges
Process samples up to 100mL with the six-place capacity Thermo Scientific™ Fiberlite™ F15-6 x 100y Fixed-Angle Rotor.
Have Questions?
Catalog NumberCapacity
750036986 x 100 mL
Catalog number 75003698
HSN 84219100
Price (SGD)
5,802.00
Special offer
Ends: 30-Jun-2026
8,288.00
Save 2,486.00 (30%)
Each
Capacity:
6 x 100 mL
Price (SGD)
5,802.00
Special offer
Ends: 30-Jun-2026
8,288.00
Save 2,486.00 (30%)
Each
Process samples up to 100mL with the six-place capacity Thermo ScientificTM FiberliteTM F15-6 x 100y Fixed-Angle Rotor.
Fiberlite Rotors:
  • Reduced run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Corrosion-and fatigue-resistant design that ensures structural integrity
  • Backed by a 15-year warranty
Specifications:
  • Capacity: 6 x 100mL
  • Max. speed/RCF: 15,000rpm/24,652 x g†
Ordering Information:
Tube Adapters for F15-6x100 Rotor sold separately. O-ring Refresher Kit (021-069-031) is also available; contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ Legend™ X1, Sorvall Legend XT, Sorvall ST 16, Sorvall ST 40, Multifuge™ X1, Multifuge™ X3, Megafuge 16, Megafuge 40, SL 16, SL 40 Series, Eppendorf™ 5810/5804 Series and Thermo Scientific Nalgene™ bottles.
Recommended for:
Pelleting bacteria or yeast cells and for purifying nucleic acids and cells.
Note:
†Performance may vary by centrifuge model
Specifications
DescriptionFiberlite F15-6 x 100y Fixed Angle Rotor
TypeFixed Angle Rotor
Capacity6 x 100 mL
Max. RCF24,652 x g
Max. Speed15,000 rpm
Radius (Metric) Min.2.5 cm
K Factor1536
Max. Tube Length105 mm
Max. Tube Diameter38 mm
For Use With (Equipment)Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Multifuge X3/X3F, Multifuge X1 and SL 40F Centrifuges
Angle25°
BiocontainmentYes
Unit SizeEach