TX-750 Rectangular Bucket Adapters
TX-750 Rectangular Bucket Adapters
Actual product may vary
TX-750 Rectangular Bucket Adapters
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500368550 mL Conical Tubes
7500368415 mL Conical Tubes
Catalog number 75003685
Price (SGD)
1,245.00
EA
Add to cart
Capacity:
50 mL Conical Tubes
Price (SGD)
1,245.00
EA
Add to cart
Use Thermo Scientific™ TX-750 Rectangular Bucket Adapters with TX-750 Rectangular Rotor Buckets.
Ordering Information:
Order rectangular buckets (75003614), Thermo Scientific™ ClickSeal™ Biocontainment Lids (75003615) and TX-750 Rotor (75003607) separately. Contact your sales representative for more information.
Specifications
TypeTube Adapter
Capacity50 mL Conical Tubes
Unit SizeEA