BIOLiner™ Rotor Bucket Adapters
BIOLiner™ Rotor Bucket Adapters
Actual product may vary
BIOLiner™ Rotor Bucket Adapters
Combine these rotor bucket adapters with the BIOLiner Rotor Bucket for a complete system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity
7500367440 x 50 mL Conical Tubes
750036715/7 mL Blood Collection Tube
7500367210 mL Blood Collection Tube
Catalog number 75003674
Price (SGD)
1,781.00
EA
Add to cart
Capacity:
40 x 50 mL Conical Tubes
Price (SGD)
1,781.00
EA
Add to cart
Combine Thermo Scientific™ BIOLiner™ Rotor Bucket Adapters with the BIOLiner Rotor Bucket for a complete system.
Ordering Information:
Individual adapters and bottle/adapter packages are offered.
  • Capacity range: 3L to 980mL
  • Maximum tube dimensions (O.D. x L): 98 × 124mm
Includes:
75-003-692 includes 750mL flat-bottom bottle with all round adapters for the TX-750 round bucket and four 500mL Corning™ conical bottle adapters.
Specifications
TypeAdapter
Tube Type50 mL Conical Bottle
Capacity40 x 50 mL Conical Tubes
Max. Tube Diameter30 mm
Max. Tube Length120 mm
No. per Pack4
Unit SizeEA