Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Actual product may vary
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor
Integrate these microplate carriers with the microplate swinging bucket rotor.
Have Questions?
Catalog number 75003625
Price (SGD)
2,824.00
EA
Add to cart
Price (SGD)
2,824.00
EA
Add to cart
Integrate Thermo Scientific™ Microplate Carriers for M-20 Microplate Swinging-Bucket Rotor to add flexibility to the swinging-bucket rotor.
Includes:
Sealed carriers include trays and lids (set of 2). Replacement O-rings for lids are offered as a set of four, with grease.
Specifications
TypeMicroplate Carrier
DescriptionSealed Microplate Carriers
Certifications/ComplianceBiocontainment certified by CAMR™, HPA, Porton Down, U.K.
Unit SizeEA