Search Fisher Scientific
| Catalog Number | Description |
|---|---|
| 75003306 | 42 x 5/7 mL Adapter |
| 75003303 | 8 x 50 mL Adapter |
| 75003304 | 18 x 15 mL Adapter |
| 75003305 | 33 x 10/12 mL Adapter |
| 75003308 | 400 mL Bottle/Centrilab Adapter |
| 75003300 | Cross |
| 75003301 | Buckets |
| 75003302 | Biocontainment Lids |
| 75003307 | Microplate Adapter |