H-Flex 1 Swinging Bucket Rotor and Accessories
Actual product may vary
H-Flex 1 Swinging Bucket Rotor and Accessories
Spin more with less! High capacity and flexibility—all in a small footprint.
Have Questions?
Catalog NumberDescription
75003301Buckets
Catalog number 75003301
HSN 84219100
Price (SGD)
1,423.00
Special offer
Ends: 30-Jun-2026
1,674.00
Save 251.00 (15%)
Each
Description:
Buckets
Price (SGD)
1,423.00
Special offer
Ends: 30-Jun-2026
1,674.00
Save 251.00 (15%)
Each
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
  • Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
  • Spins blood tubes, conical tubes and microplates without changing rotor or buckets
  • Get results faster without the need to change hardware
  • Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
DescriptionBuckets
MaterialForged/Machined Aluminum with Anodized Coating
TypeSwinging Bucket Rotor
CapacitySee Individual Adapter Capacities
Max. Speed4297
For Use With (Equipment)Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series
Angle90°
Certifications/ComplianceBiocontainment Certified by CAMR™, HPA, Porton Down, UK
BiocontainmentYes, when used with biocontainment lids
AutoclavableAutoclavable
QuantitySet of 2
Unit SizeEach