TCF-20 Continuous Flow Rotor
TCF-20 Continuous Flow Rotor
Actual product may vary
TCF-20 Continuous Flow Rotor
Perform bioprocessing or separation of large volumes with very low solids content.
Have Questions?
Catalog NumberCapacity
750030121350 mL
Catalog number 75003012
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
1350 mL
Perform bioprocessing or separation of large volumes with very low solids content using the Thermo Scientific™ TCF-20 Continuous Flow Rotor. Rotor innovations—from Thermo Scientific™ Auto-Lock™ rotor exchange to Thermo Scientific™ Auto-ID™ instant rotor identification and speed handles on each rotor lid—shorten run set-up time while providing peace-of-mind the rotor is secure.
Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Auto-ID Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionTCF-20 Continuous Flow Rotor
MaterialTitanium
TypeContinuous Flow Rotor
Capacity1350 mL
Max. RCF42,913 x g
Max. Speed20,000 rpm
Radius (Metric)96 mm
Radius (Metric) Min.26 mm
K Factor826
Net Weight17.2 lb. (7.8 kg)
For Use With (Equipment)Sorvall LYNX 6000 Superspeed Centrifuge
BiocontainmentYes
Unit SizeEA