TH13-6 x 50 Swinging Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
TH13-6 x 50 Swinging Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Actual product may vary
TH13-6 x 50 Swinging Bucket Rotor for LYNX 4000 and 6000 Superspeed Centrifuges
Achieve gradient separations or pelleting directly to the bottom of the tube with these rotors, designed for round or conical tubes.
Have Questions?
Catalog NumberCapacity
750030106 x 50 mL
Catalog number 75003010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
6 x 50 mL
Achieve gradient separations or pelleting directly to the bottom of the tube with the Thermo Scientific™ TH13-6 x 50mL Swinging-Bucket Rotor, designed for round or conical tubes.
Thermo Scientific™ Auto-Lock Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
Specifications
DescriptionTH13-6x50 Swinging Bucket Rotor
MaterialAluminum with Titanium Buckets
TypeSwinging Bucket Rotor
Capacity6 x 50 mL
Max. RCF30,314 x g
Max. Speed13,100 rpm
Radius (Metric)107 mm
Radius (Metric) Min.33 mm
K Factor1,503
Max. Tube Length104 mm
Max. Tube Diameter29 mm
Net Weight15.8 lb. (7.2 kg)
For Use With (Equipment)Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle90°
BiocontainmentYes
Unit SizeEA