CW3 Cell Washer Rotors
Actual product may vary
CW3 Cell Washer Rotors
CW3 Cell Washer Rotors
CW3 Cell Washer Rotors
Thorough, reproducible blood cell washing of 12 or 24 tubes per run with Thermo Scientific™ CW3 cell washer rotors.
Have Questions?
Catalog NumberCapacity
7500002024 Tubes
Catalog number 75000020
HSN 84219100
Price (SGD)
-
Capacity:
24 Tubes
Meet your protocol requirements with the flexibility to run 12 or 24 samples and finish washing in as little as 3 minutes with Thermo Scientific™ CW3 cell washer rotors.
The Thermo Scientific™ CW3 Cell Washer has been discontinued.
Optimize pelleting performance
  • For washing, tubes swing at a fixed positive angle to sediment cells rapidly at the bottom of the tube
  • For decanting, the rotor holds tubes at a slightly negative angle and the saline is decanted centrifugally
Designed for safe operation
  • Saline distributor mounted directly on top of the rotor to reduce risk of cross-contamination
  • Easy to remove for cleaning convenience
  • Stainless steel rotor body advances system durability
Specifications
Description24-place Rotor Assembly with Distributor
MaterialStainless Steel
TypeFixed Angle Rotor
Capacity24 Tubes
Max. RCF1,180 x g (Centrifuge), 190 x g (Wash)
Max. Speed3,000 rpm (Centrifuge), 200 rpm (Wash), 350 to 500 rpm (Decant)
Radius (Metric)117 mm
Max. Tube Length75 mm
Max. Tube Diameter12 or 10 mm
For Use With (Equipment)CW3 Cell Washer
Unit SizeEach