Check Order Status
Type IAA Fiberglass Step Ladder with EDGE Bracing System
Actual product may vary
Thermo Scientific
Type IAA Fiberglass Step Ladder with EDGE Bracing System
Features extra heavy duty aluminum holster tops, internal spreaders, the EDGE bracing system, and external rail shields for long lasting durability.
Have Questions?
Change view
Catalog Number
Length (English)
7403
36 in.
7404
2 Options
Catalog number
7403
Price (SGD)
29,512.00
EA
Add to cart
Length (English):
36 in.
Price (SGD)
29,512.00
EA
Add to cart
Extra heavy duty aluminum HolsterTop™ with thick side plates.
Back-up plates reinforce all top connectors
Shoulder bolt and lock nut on top hinge, 4 solid aluminum rivets attach top to each front rail
Slip-resistant Traction-Tred™ steps are double riveted and have extra wide knee braces
Full set of rear horizontals are knee braced and spaced one per foot
Extra heavy duty internal aluminum spreaders
Aluminum external rail shield helps protect against abrasion damage
GLASMARK™ Violator stripes on rails allow ladder to be more visible
Slip-resistant foot pads attached to the EDGE™ structure
EDGE™ bracing system helps to decrease damage to rail
Every rivet backed up by metal, metal plates or washers to protect rails
Specifications
Certifications/Compliance
ANSI-A14-2-2007, OSHA
Dimensions (L x W x H)
36 x 19.75 x 7.2 in.
Height (English)
6 ft.
Length (English)
36 in.
Model
7400 Series
Quantity
1 Ea.
Width (English)
19.75 in.
Load Capacity (English)
375 lb.
Load Capacity (Metric)
170 kg
Material
Fiberglass
Product Type
Folding Step Ladder