Nalgene™ Syringe Filters
Nalgene™ Syringe Filters
Actual product may vary
Nalgene™ Syringe Filters
Filter sample volumes of 10 to 100mL with these sterile syringe filters featuring a variety of membranes and pore sizes for most laboratory applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeDiameter (Metric)Material (Membrane)Sterility
720-13200.2 μm13 mmPESSterile
FIS09-740-1000.2 μm13 mmPESSterile
FKS153423780.2 μm13 mmPESSterile
723-25200.2 μm25 mmSFCASterile
FIS09-740-1060.45 μm25 mmSFCASterile
723-25800.8 μm25 mmCellulose AcetateSterile
725-25450.45 μm25 mmPESSterile
FIS09-740-34A0.2 μm4 mmCellulose AcetateNon-sterile
Catalog number 720-1320
Price (SGD)
538.00
CS
Add to cart
Pore Size:
0.2 μm
Diameter (Metric):
13 mm
Material (Membrane):
PES
Sterility:
Sterile
Price (SGD)
538.00
CS
Add to cart
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Key Features
  • 10 to 100mL sample size
  • Inlet: Female Luer-Lok™
  • Outlet: Male Luer slip
  • Polypropylene housing with ID color rings
  • Pore size and membrane type printed on unit
Polyether Sulfone (PES)
  • Highest throughput and lowest clogging
  • Low binding
  • Most commonly used general purpose membrane
Surfactant-free Cellulose Acetate (SFCA)
  • Low protein binding, lower extractables than standard CA
  • Good for cell culture fluids and cellular solutions
  • Polypropylene housings with blue color rings
Cellulose Acetate (CA)
  • Some protein binding and extractables
  • For sterile filtration and clarification of: buffers, aqueous solutions and reagents
  • Polypropylene housings with red color rings
Nylon
  • Naturally hydrophilic, low extractables, some protein binding
  • Useful for alcohols, cryoprotectants such as DMSO and with aqueous/biological and weak solvents
  • Polypropylene housings with pink color rings
PTFE
  • Naturally hydrophobic
  • Excellent for HPLC/GC solvent filtration. Ideal for sterile venting of air or other gases during filtration
  • Polypropylene housings with white color rings
Specifications
Certifications/ComplianceCertified sterile, non-cytotoxic and non-pyrogenic
Filter TypeMembrane Filter
Material (Membrane)PES
Pore Size0.2 μm
SterilitySterile
For Use With (Application)Sterile filtration and clarification of cell culture media, buffers and additives
Housing MaterialPolypropylene
Quantity100/Pk.
TypeSyringe Filter
Volume (Metric)2 to 10 mL
Diameter (Metric)13 mm
Unit SizeCS
Frequently asked questions (FAQs)
Are Nalgene Sterile Syringe Filters packed individually or in bulk?
The Nalgene Sterile Syringe Filters are individually blister-packed. The Nalgene non-Sterile Syringe Filters are bulk packed.