DNA AWAY™ Surface Decontaminant
Actual product may vary
DNA AWAY™ Surface Decontaminant
Eliminate unwanted DNA and DNase from glassware and plasticware without affecting subsequent DNA samples. This surface decontaminant degrades DNA more quickly and effectively than autoclaving.
Have Questions?
Catalog NumberDescription
7010JPDNA Surface Decontaminant
Catalog number 7010JP
HSN 34029059
Price (SGD)
48.00
EA
Description:
DNA Surface Decontaminant
Price (SGD)
48.00
EA
Eliminate unwanted DNA and DNase from glassware and plastic ware without affecting subsequent DNA samples. Thermo Scientific™ DNA AWAY™ Surface Decontaminant degrades DNA more quickly and effectively than autoclaving.
  • Suitable for gel boxes, pipettes, benchtops, thermal cyclers, or other apparatus
  • Use before performing PCR or working with DNA for a DNA-free work area
  • Simply apply the ready-to-use formula to the surface to decontaminate, then wipe dry or rinse clean
Specifications
pH11
CompositionSodium Hydroxide with Water
DescriptionDNA Surface Decontaminant
Volume (English)8.5 oz.
Volume (Metric)250 mL
Unit SizeEA