Search Fisher Scientific
| Catalog Number | Volume (Metric) | Container Type |
|---|---|---|
| 7003 | 1 L | Bottle |
| 7000 | 250 mL | Bottle |
| FIS21-236-21 | 250 mL | Bottle |
| 7002 | 475 mL | Spray Bottle |
| FIS21-402-178 | 475 mL | Spray Bottle |
| FKS10666421 | 475 mL | Spray Bottle |
| 7005-11 | 4 L | Bottle |
| FIS14-754-34 | 4 L | Bottle |
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.