RNase AWAY™ Surface Decontaminant
Actual product may vary
RNase AWAY™ Surface Decontaminant
Eliminate RNase and DNA from laboratory surfaces. This product reduces dependency on carcinogenic DEPC treatments and saves time spent baking glassware.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Container Type
7002475 mLSpray Bottle
7000250 mLBottle
FIS21-236-21250 mLBottle
FIS21-402-178475 mLSpray Bottle
FKS10666421475 mLSpray Bottle
70031 LBottle
7005-114 LBottle
FIS14-754-344 LBottle
Catalog number 7002
Price (SGD)
48.00
Special offer
Ends: 30-Jun-2026
59.00
Save 11.00 (19%)
Each
Volume (Metric):
475 mL
Container Type:
Spray Bottle
Price (SGD)
48.00
Special offer
Ends: 30-Jun-2026
59.00
Save 11.00 (19%)
Each
Eliminate RNase and DNA from laboratory surfaces. Thermo Scientific™ RNase AWAY™ is ideal for decontaminating apparatus, bench-tops, glassware and plasticware. It reduces dependency on carcinogenic DEPC treatments and saves time needed to bake glassware.
  • Use on pipettes, gel boxes, or RNA or DNA prep areas
  • Leaves no residue to interfere with gel polymerization or staining
  • Chemically stable and nonabrasive
  • Contains no strong acids
Specifications
pH11
CompositionSodium Hydroxide with Water
Container TypeSpray Bottle
Volume (English)16 oz.
Volume (Metric)475 mL
Unit SizeEach
Frequently asked questions (FAQs)
Do you have a Technical data sheet (TDS) for RNase AWAY Surface Decontaminant?
We have a safety data sheet (SDS) and COA for RNase AWAY Surface Decontaminant. Visit our Documents and Certificates page to search for and request these documents.