Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Actual product may vary
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray with Cover
Reduce breakage and chipping of glass pipette tips with the stackable and lightweight Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray.
Have Questions?
Catalog NumberDimensions (L x W x H)
6910-061818 x 6 x 2.6 in.
Catalog number 6910-0618
Price (SGD)
471.00
CS
Add to cart
Dimensions (L x W x H):
18 x 6 x 2.6 in.
Price (SGD)
471.00
CS
Add to cart
Reduce breakage and chipping of glass pipette tips with the Thermo Scientific™ Nalgene™ Polypropylene Pipet/Instrument Sterilizing Tray. Horizontal design allows pipettes to be fully immersed in disinfectant. Rigid plastic tray is stackable, lightweight, quiet and less expensive than stainless steel. Cover fits into groove of pan to minimize spillage of hazardous materials.
  • Holds pipets up to 40cm long in a stackable, lightweight alternative to stainless steel
  • Can also be used to sterilize small labware and instruments—resists aggressive disinfectants
  • Has groove for cover to minimize spillage of hazardous materials and a hand grip on the cover
  • Autoclavable
Compliance:
  • Meets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens.
  • Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of pipets are required before cleaning and disposal
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Certifications/ComplianceMeets OSHA Standard 29CFR1910.1030 for use as protection against bloodborne pathogens. Conforms to NIH and Federal Register guidelines for biohazardous and recombinant DNA research where disinfecting and subsequent autoclaving of
Dimensions (L x W x H)18 x 6 x 2.6 in.
DescriptionSterilizing tray with rigid plastic construction
Height (Metric)6.7 cm
Length (Metric)46 cm
MaterialPolypropylene
TypeSterilizing Tray
Width (Metric)15 cm
For Use WithHolds pipets up to 40cm long
Length (English)18 in.
No. per Pack1
Height (English)2.6 in.
No. per Case6
ShapeRectangular
Width (English)6 in.