Model Plus Animal Cell
Model Plus Animal Cell
Actual product may vary
Model Plus Animal Cell
Colorful teaching model designed specifically for hands-on use.
Have Questions?
Catalog NumberGrade
6907-21 to 8
Catalog number 6907-2
Price (SGD)
-
Grade:
1 to 8
  • Five inquiry-based activities cover cell structure and function, osmosis, eukaryotes, bacteria and more
  • Suitable for elementary through middle school
  • Resilient, nontoxic EVA-foam construction withstands years of active investigation
  • Dimensions: 11L x 11W x 14.5 in.H (27.9 x 27.9 x 36.8cm)
Specifications
Product TypeCell Model Plus, Animal
Dimensions (L x W x H)11 x 11 x 14.5 in. (27.9 x 27.9 x 36.8 cm)
For Use With (Application)Colorful teaching model designed specifically for hands-on use
Grade1 to 8
Height (English)14.5 in.
Height (Metric)36.8 cm
Includes5 Inquiry-based lab activities, reproducible worksheets, complete background information, cross-curricular extension activities, assessment ideas and more
Length (English)11 in.
Length (Metric)27.9 cm
MaterialResilient, Non-toxic EVA Foam
Width (English)11 in.
Width (Metric)27.9 cm
Unit SizeEA