Nalgene™ Autoclavable Polypropylene Pans
Actual product may vary
Thermo Scientific
Nalgene™ Autoclavable Polypropylene Pans
Repeatedly autoclave instruments and labware with these autoclavable polypropylene pans, featuring heavy-duty construction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
6902-50005 L
6902-10002 L
FIS13-361-202 L
FIS13-361-152.8 L
6902-30003.1 L
FIS13-361-103.1 L
FIS13-361-255 L
Catalog number 6902-5000
HSN 39239090
Price (SGD)
76.00
EA
Capacity (Metric):
5 L
Price (SGD)
76.00
EA
Repeatedly autoclave instruments and labware with Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Pans, featuring heavy-duty construction, extra strong rims and rounded corners for easy cleaning.
  • Tapered for nesting to save space
  • Rolled edges provide extra stiffness for carrying heavy contents
  • Heavy wall construction will withstand rugged use and repeated autoclaving
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Capacity (Metric)5 L
Dimensions (L x W x H)12.75 x 10.125 x 4.25 in.
DescriptionPolypropylene Autoclavable Pan
Height (Metric)108 mm
Length (Metric)324 mm
MaterialPolypropylene
TypePan
Width (Metric)257 mm
Length (English)12.75 in.
No. per Pack1
Height (English)4.25 in.
ShapeRectangular
Width (English)10.125 in.