LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
Actual product may vary
LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
LSE™ Multiposition Digital Vortexer
Corning™ LSE Multipostion Digital Vortexer is a multipurpose shaker dedicated to a variety of aggressive shaking and mixing applications.
Have Questions?
Catalog NumberVoltage
6709115 V
Catalog number 6709
Price (SGD)
1,908.00
EA
Add to cart
Voltage:
115 V
Price (SGD)
1,908.00
EA
Add to cart
The Corning Digital Vortexer is a multipurpose shaker dedicated to a variety of shaking and mixing applications. Three easily removable and interchangeable platforms accommodate many different sample containers such as flasks, bottles, tube racks, plates, boxes, and dishes. The clamp platform has spring loaded retaining bars that adjust to fit vessels of different shapes and sizes. The microplate platform is designed to securely hold up to four microplates. Operation of the vortexer can be timed or continuous, while the speed is variable. The broad speed range allows the vortexer to be used for gentle mixing as well as vigorous agitation. A powerful motor, and a chemically resistant and easy to clean surface makes the equipment a perfect choice for years of trouble-free use.
Features and Benefits
  • Three versatile platform options (available separately)
  • Compact design with easy-access control panel
  • Non-slip rubber mat to keep containers in place
  • Safe for cold room or incubator use
  • Five-year warranty
Specifications
Load Bearing Capacity (Metric)2 kg
Speed Range100 to 1200 rpm
Voltage115 V
Temperature Range (Metric)4°C to 65°C
Product TypeVortexer
Load Bearing Capacity (English)4.4 lb.
Frequency50/60 Hz
Dimensions (L x W x H)33.7 x 26.2 x 13 cm (13.3 x 10.3 x 5.1 in.)
Shipping Weight (English)16 lb.
Shipping Weight (Metric)7.3 kg
DescriptionRequires a platform
For Use With (Application)General Purpose Mixing
Timer Range0 to 99 minutes or continuous
Warranty and Service Offering5 Years
Unit SizeEA