Nalgene™ Acrylic Benchtop Beta Radiation Shield
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Actual product may vary
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Nalgene™ Acrylic Benchtop Beta Radiation Shield
Protect lab workers with these benchtop radiation shields, specifically designed for use during procedures which require the handling of beta-emitting isotopes.
Have Questions?
Catalog NumberDepth (Metric) InteriorWidth (Metric) InteriorHeight (Metric) Interior
6700-181230 cm30 cm46 cm
Catalog number 6700-1812
Price (SGD)
336.00
CS
Add to cart
Depth (Metric) Interior:
30 cm
Width (Metric) Interior:
30 cm
Height (Metric) Interior:
46 cm
Price (SGD)
336.00
CS
Add to cart
Protect lab workers with the Thermo Scientific™ Nalgene™ Acrylic Benchtop Beta Radiation Shield, specifically designed for use on the laboratory benchtop during procedures which require the handling of β-emitting isotopes. Made of 3/8 in. (9mm) thick acrylic, shields are transparent and free-standing with an angled top that allows contents of tubes located behind the shield to be viewed from the top. Available in two sizes. Meets OSHA Standard 29CFR Part 1910.1030 for use as protection against bloodborne pathogens.
  • Designed for use on the laboratory benchtop during procedures that require the handling of β-emitting isotopes.
  • Made of 3/8 in. (9mm) thick acrylic — stops β-emitting radioactive isotopes
  • Transparent with an angled top that allows contents of tubes located behind the shield to be viewed from the top
  • 12 in. (305mm) large flat work surface that minimizes the risk of hot spots on the benchtop
Includes:
Radiation labels
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
DescriptionAngled-top, Beta Radiation Shield
MaterialLDPE, LDPE
Depth (English) Interior12 in.
AutoclavableNot Autoclavable
Capacity (English)52 oz.
Depth (Metric) Interior30 cm
Capacity (Metric)1538 mL
Closure TypeTape and Wire
ColorClear with White Block
DisposableSingle-use
For Use With (Application)Collect, Store, and Transport Samples
Height (English) Interior18 in.
For Use With (Equipment)Stomacher, Blender, Homogenizer
IncludesBag
Height (Metric) Interior46 cm
Thickness (English)0.37 in.
SterilitySterile
Thickness (Metric)0.102 mm
Tubing StyleContinuous perimeter seal with tape and wire closure, full-page filter
Fill Volume52 oz.
Width (English) Interior12 in.
Width (Metric) Interior30 cm
Latex FreeYes
Item DescriptionShield Benchtop Beta Acrylic 457 x 305 x 305 mm
Certifications/ComplianceBoth shields meet OSHA Standard 29CFR Part 1910.1030 for use as protection against bloodborne pathogens.
No. per Pack500/Pk.
Outer PackagingCardboard
Product LineBlender Bag
Protective Inner PackagingPlastic
Sterilization MethodEthylene Oxide Gas
Temperature Range (English)-320°F to +180°F
Temperature Range (Metric)-196°C to +82°C
Thickness (English)4 mil
Dimensions (L x W)33 x 18.5 cm (13 x 7 in.)
Length (English)13 in.
Length (Metric)33 cm
Width (English)7 in.
Width (Metric)18.5 cm
Product TypeBlender Bag (Round Bottom)
Unit SizeCS