Nalgene™ Safety Waste System
Nalgene™ Safety Waste System
Actual product may vary
Nalgene™ Safety Waste System
Nalgene™ Safety Waste System
Nalgene™ Safety Waste System
Nalgene™ Safety Waste System
Nalgene™ Safety Waste System
Minimize accidental spills and volatile emissions with this chemical- and break-resistant safety waste system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeDiameter (Metric) OpeningMaterial
6379-001010 L83B25 cmFLPE
6379-00044 L38-43014 cmHDPE
FIS10-437-3610 L83B25 cmFLPE
Catalog number 6379-0010
Price (SGD)
248.00
CS
Add to cart
Capacity (Metric):
10 L
Closure Size:
83B
Diameter (Metric) Opening:
25 cm
Material:
FLPE
Price (SGD)
248.00
CS
Add to cart
Minimize accidental spills and volatile emissions with the chemical- and break-resistant Thermo Scientific™ Nalgene™ Safety Waste System, designed for temporary storage of solvent, chemical and biological wastes. Safety waste funnel has an Easy Snap safety latch to minimize accidental spills and volatile emissions. Funnel easily unscrews from funnel adapter for cleaning and quicker access. Closure/adapter includes a vent that releases pressure when pouring, and reduces emmisions during use. Includes a Nalgene collection container.
Features:
  • Large-diameter funnel has attached cover with Easy Snap safety latch to minimize accidental spills and volatile emmissions
  • Funnel easily unscrews from polypropylene closure/funnel adapter for cleaning and quicker access
  • Closure/adapter includes vent with polypropylene/PTFE vent plug that releases pressure during pouring, and prevents emissions during use
  • Adapter can also connect to external syringe filter
  • Removable polypropylene screen insert fits at bottom of funnel to trap large particles such as stir bars
  • Not resistant to long-term UV light exposure
  • Includes either a 4L HDPE Nalgene bottle or 10L FLPE Nalgene Carboy
  • 4L system include a 5-1/2 in. (14cm) safety waste funnel and 4L HDPE Nalgene bottle and closure. Recommended for acid, base and alcohol wastes.
  • 10L system includes a 10 in. (25cm) safety waste funnel and 10L fluorinated HDPE carboy with closure. Recommended for solvent wastes.
Ordering Information:
Secondary containers (sold separately) should be used with Safety Waste Systems during transport, transfer or storage.
Includes:
1.125 gal. (4L) HDPE or 2.625 gal. (10L) fluorinated HDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insert.
Note:
These systems are not safety cans as defined by OSHA guidelines and are designed for temporary waste disposal only. Waste should be disposed of prior to 90 days of initial waste accumulation.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Closure Size83B
Capacity (Metric)10 L
DescriptionSafety Waste Systems; Capacity: 10L
Diameter (English) Outer10 in.
Capacity (English)2.625 gal.
Length (Metric)27 cm
Diameter (Metric) Opening25 cm
Includes2.625 gal. (10L) fluorinated HDPE container, removable funnel with lid, closure/funnel adapter, filter, and screen insert.
For Use With (Application)For temporary waste containment
Length (English)10.625 in.
MaterialFLPE
Product TypeSafety Waste System
Unit SizeCS