Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Actual product may vary
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Simultaneously maintain safety and a clear field of vision by using this safety splash shield. Provides protection from splashes, aerosols and flying debris.
Have Questions?
Catalog NumberDescriptionLength (English)Width (English)
6356-0001Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm12 in.8.5 in.
Catalog number 6356-0001
Price (SGD)
205.00
EA
Add to cart
Description:
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm
Length (English):
12 in.
Width (English):
8.5 in.
Price (SGD)
205.00
EA
Add to cart
Simultaneously maintain safety and a clear field of vision by using the Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield. This shield attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. This durable, scratch-resistant 1/8 in. (3mm) thick polycarbonate shield protects the user and features an easy to maneuver arm. Meets OSHA 29CFR1910.1030 standard.
  • Flexible 14 inch arm makes it easy to maneuver shield over samples
  • Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
  • Provides UV protection from 200-360nm for added versatility
  • Biohazard/Transparent
Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 
Specifications
Thickness (English)0.12 in.
Thickness (Metric)3 mm
Width (English)8.5 in.
Width (Metric)21.5 cm
Certifications/ComplianceMeets OSHA 29CFR1910.1030 standard
DescriptionSafety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm
Lens TintClear
Product TypeSafety Splash Shield
Length (English)12 in.
No. per Case4/Cs.
No. per Pack1 Ea.
MaterialPolycarbonate (Lens)
Lens MaterialPolycarbonate
UV ProtectionYes
Unit SizeEA