HOSTESS™ C-Fold Green Hand Towels
HOSTESS™ C-Fold Green Hand Towels
Actual product may vary
HOSTESS™ C-Fold Green Hand Towels
Dispenses one at a time to prevent the spread of germs. Kimberly-Clark™ HOSTESS™ C-Fold Green Hand Towels dry your hands hygienically and effectively.
Have Questions?
Catalog NumberDimensions (L x W)
601025 x 33 cm
Catalog number 6010
Price (SGD)
-
Dimensions (L x W):
25 x 33 cm
  • Helps reduce costs and waste
  • Ideal for busy washrooms (schools and other facilities)
  • Environmentally friendly; carries the EU Ecolabel
Specifications
Product TypeHand Towel
Length (English)23.4 in.
Length (Metric)25 cm
Width (Metric)33 cm
MaterialCrepe
Basis Weight12kg (25.036 lbs.)
Certifications/ComplianceISO 14001, ISO 9001
ColorGreen
Dimensions (L x W)25 x 33 cm
FormatC-Fold
Packaging Quantity20 x 180 Sheets
Ply1-ply
Unit SizeEach