Air Filter Kits
Air Filter Kits
Actual product may vary
Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Description
598000000009ThermoFlex™ 3500 or 5000 Recirculating ChillerAir and fluid filter kit (35 micron)
598000000005ThermoFlex™ 900 or 1400 Recirculating ChillerAir and fluid filter kit (35 micron)
Catalog number 598000000009
Price (SGD)
-
For Use With (Equipment):
ThermoFlex™ 3500 or 5000 Recirculating Chiller
Description:
Air and fluid filter kit (35 micron)
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific Neslab HX, CFT or M series recirculating chillers.
Specifications
DescriptionAir and fluid filter kit (35 micron)
For Use With (Equipment)ThermoFlex™ 3500 or 5000 Recirculating Chiller
TypeAir and Fluid Filter
Unit SizeEA