Search Thermo Fisher Scientific
Catalog Number | Positions | Shape |
---|---|---|
5974-4015 | 8 | Round |
5974-0404 | 16 | Square |
FIS14-809-166 | 16 | Square |
5974-1015 | 20 | Round |
FIS14-809-166A | 20 | Round |
FKS10488692 | 20 | Round |
FKS10613991 | 8 | Round |
Thaw, cool and incubate samples at elevated or reduced temperatures with Thermo Scientific™ Nalgene™ Polypropylene Floating Microtube Racks. Especially useful in genetic cloning, microbiology and immunology labs, these racks offer excellent temperature resistance from -70° to +100°C (-94° to +212°F). Tubes are supported in the rack below the lip. Square or round racks have molded-in letters and numbers at each hole for easy sample identification.
Features:
Note:
Racks are not autoclavable.
Nalgene reusable polypropylene(PP) products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. These racks and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged. And at end of product life, please wash and recycle your Nalgene racks if services are available in your community.