Check Order Status
Lindberg/Blue M™ Tube Furnace Adapters/Sleeves
Lindberg/Blue M™ Tube Furnace Adapters/Sleeves
Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
Have Questions?
Catalog Number
Description
For Use With (Equipment)
59524
2.5 in. Adapter
HTF55322, HTF55342, HTF55347 Tube Furnaces
Catalog number
59524
Price (SGD)
521.00
EA
Add to cart
Description:
2.5 in. Adapter
For Use With (Equipment):
HTF55322, HTF55342, HTF55347 Tube Furnaces
Price (SGD)
521.00
EA
Add to cart
Use Thermo Scientific™ Lindberg/Blue M™ Tube Furnace Adapters/Sleeves with Lindberg/Blue M Tube Furnaces.
Specifications
Description
2.5 in. Adapter
For Use With (Equipment)
HTF55322, HTF55342, HTF55347 Tube Furnaces
Type
Tube Furnace Adapter
Diameter (Metric) Process Tube
63.5 mm