Nalgene™ Petri and Bioassay Dish Racks
Nalgene™ Petri and Bioassay Dish Racks
Actual product may vary
Nalgene™ Petri and Bioassay Dish Racks
Secure glass and plastic dishes for safe transport with these petri and bioassay dish racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsFor Use With (Equipment)Autoclavable
5920-0060Petri Dishes3.93 in. (100mm) dishAutoclavable
FIS08-759-10APetri Dishes3.93 in. (100mm) dishAutoclavable
FIS08-759-10BPetri Dishes3.93 in. (100mm) dishNot Autoclavable
Catalog number 5920-0060
Price (SGD)
396.00
CS
Add to cart
Holds:
Petri Dishes
For Use With (Equipment):
3.93 in. (100mm) dish
Autoclavable:
Autoclavable
Price (SGD)
396.00
CS
Add to cart
Secure glass and plastic dishes for safe transport with Nalgene™ Petri and Bioassay Dish Racks. Requiring only simple assembly, these clear racks permit side visibility for culture checks during incubation.
  • Clear racks permit side visibility for culture checks during incubation
  • Simple assembly required
Specifications
Product TypePetri Dish Rack
AutoclavableAutoclavable
ColorClear
HoldsPetri Dishes
MaterialPolycarbonate
For Use With (Equipment)3.93 in. (100mm) dish
No. per Case2
No. per Pack1
Diameter (Metric) Inner100 mm
Unit SizeCS