Nalgene™ Acrylic Pipet Box Holder
Nalgene™ Acrylic Pipet Box Holder
Actual product may vary
Nalgene™ Acrylic Pipet Box Holder
Nalgene™ Acrylic Pipet Box Holder
Nalgene™ Acrylic Pipet Box Holder
Nalgene™ Acrylic Pipet Box Holder
Nalgene™ Acrylic Pipet Box Holder
Store bags, boxes, pipet canisters or 1 to 100mL individually-wrapped serological pipets.
Have Questions?
Catalog NumberPositions
5810-00044
Catalog number 5810-0004
Price (SGD)
826.00
CS
Add to cart
Positions:
4
Price (SGD)
826.00
CS
Add to cart
Angled Pipet Holder prevents pipets from sliding out. Accommodates bags, boxes, pipet canisters or 1 - 100mL individually-wrapped serological pipets.
  • Clear acrylic organizers allow you to view their contents easily and keep your small lab items neatly organized and within reach
  • Can withstand brief exposure to dilute acids, and bases—Alcohols, organic solvents and strong acids may cause immediate damage
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Product TypePipet Box Holder
DescriptionPipet Holder Angled 4-place 3-1/2 in. x 16-3/8 in. x 11-1/2 in.
HoldsSerological Pipets (1 to 100 mL)
MaterialAcrylic
Positions4
ShapeRectangular
No. per Case1
QuantityCase of 1
Dimensions (L x W x H)3.5 x 16.4 x 11.5 in. (8.9 x 41.6 x 29.2cm)
Length (Metric)8.9 cm
Length (English)3.5 in.
Width (Metric)41.6 cm
Width (English)16.4 in.
Height (Metric)29.2 cm
Height (English)11.5 in.
Unit SizeCS