Nalgene™ Petri Dish
Nalgene™ Petri Dish
Actual product may vary
Nalgene™ Petri Dish
Nalgene™ Petri Dish
Nalgene™ Petri Dish
Nalgene™ Petri Dish
Shatterproof and reusable
Have Questions?
Catalog NumberDiameter (Metric)
5500-0010100 mm
Catalog number 5500-0010
Price (SGD)
583.00
CS
Add to cart
Diameter (Metric):
100 mm
Price (SGD)
583.00
CS
Add to cart
  • Resists cell adhering
  • Stackable for convenience
  • Top and bottom interlocking ridges
  • Autoclavable
Specifications
AutoclavableAutoclavable
MaterialPMP
ShapeRound
SterilitySterile
TypePetri Dish
EdgesStackable
QuantityPk, of 10
Height (Metric)15 mm
Diameter (Metric)100 mm
Unit SizeCS