Skullerz™ Dagr Safety Glasses/Sunglasses, Black Frame
Actual product may vary
Skullerz™ Dagr Safety Glasses/Sunglasses, Black Frame
Protective work safety sunglasses feature tough scratch-resistant lenses that filter 99.9% of harmful UVA, UVB and UVC rays to keep eyes safe in the sun and the blade-style design is a sleek and sporty look.
Have Questions?
Catalog NumberLens Tint
52030Smoke
Catalog number 52030
HSN 84219100
Price (SGD)
6,123.00
Special offer
Ends: 30-Jun-2026
8,747.00
Save 2,624.00 (30%)
Each
Lens Tint:
Smoke
Price (SGD)
6,123.00
Special offer
Ends: 30-Jun-2026
8,747.00
Save 2,624.00 (30%)
Each
  • Durable half frames bend and flex without breaking
  • Non-slip rubber nose piece and rubber temples keep the glasses secure and resist sweat
  • Available with a polarized lens or with anti-fogging solution
  • Polarized glasses reduce glare and eye fatigue around water, snow and in other bright environments.
  • Anti-glare glasses block the harsh reflected light from water
  • Fog-Off™ treatment won't wash off or wear off for fog-free viewing and are ideal for use with dust masks
  • Certifications and testing:
    • Impact-resistant safety glasses are tested to ANSI Z87.1+ High-Velocity Impact Protection and optical requirements
    • Tested to MIL-PRF-32432 Ballistic Fragmentation, Class 1, which is the US Military Combat Eye Protection System Performance Specification for eyewear. To pass the military standard, safety glasses must withstand a steel projectile shot at 660 ft / 201m per second which is 4x faster than the Z87+ standard. Lenses must not puncture, crack or shatter. Frames must retain the lenses and not break.
  • Ideal for outdoor work like construction, carpentry/woodworking or landscaping or for outdoor activities like boating, skiing, fishing, hunting/shooting, and sports.
  • Available in a variety of lens options ideally suited for a variety of tasks and light environments
Specifications
Lens CoatingImpact-resistant, Scratch-resistant, UV-treated
Certifications/ComplianceANSI Z87.1+, CSA Z94.3 + and MIL-PRF 32432 Ballistic Impact
Lens TintSmoke
For Use With (Application)Construction/Trades, Oil and Gas, Mining, Maintenance, Assembly/Fabrication, Manufacturing, Police/Tactical
Frame ColorBlack
No. per Case12/Cs.
Unit SizeEach