Heraguard™ ECO Clean Bench Accessories
Heraguard™ ECO Clean Bench Accessories
Actual product may vary
Heraguard™ ECO Clean Bench Accessories
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeDescriptionFor Use With (Equipment)
51901075Work Surface WrapsStainless Steel Work SurfaceHeraguard ECO 1.2
50044678Media ValvesVacuum tap G3/8 connection (grey) with long neck for installation in side wallsHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95; Herasafe 2030i 0.9, 1.2, 1.5, 1.8; Maxisafe 2030i 0.9, 1.2, 1.5, 1.8
51901073UV OptionsUV Option, factory installed, comes with UV-resistant Night CoverHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
51901077Work Surface WrapsStainless Steel Work SurfaceHeraguard ECO 1.8
51901072Side Window ModificationsRight hand glass with metal sheet and 3 media valve holesHeraguard ECO 0.9, 1.2, 1.5, 1.8, 1.8/95
Catalog number 51901075
Price (SGD)
1,049.00
EA
Add to cart
Product Type:
Work Surface Wraps
Description:
Stainless Steel Work Surface
For Use With (Equipment):
Heraguard ECO 1.2
Price (SGD)
1,049.00
EA
Add to cart
Integrate Thermo Scientific™ Heraguard™ ECO Clean Bench Accessories with the ECO clean bench to increase the flexibility and enhance operations. Options include media valves, solenoid valve, stainless-steel work surface wraps, side window modifications, UV and UV night covers, windscreen protection kits and additional accessories.
†Due to DC motors, Heraguard ECO clean benches use less energy to operate so there is less heat emitted into the lab than traditional AC motor clean benches. Based on data from internal testing and The University of Michigan Field Study, published in American Biotechnology Laboratory.
Specifications
For Use With (Equipment)Heraguard ECO 1.2
Depth (Metric)580 mm
DescriptionStainless Steel Work Surface
Width (Metric)1200 mm
Product TypeWork Surface Wraps
Configuration OptionsFactory Installed
Unit SizeEA