Thermo Scientific
Thermo Scientific™ QSP™ Natural Screw Cap Micro Tubes
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection great with biologics, sample preparation, storage and transportation.
Have Questions?
Catalog NumberClean ClaimsAutoclavableSterility
517-QRNase/DNase, DNA, pyrogen and endotoxin freeNot AutoclavableNon-sterile
Catalog number 517-Q
HSN 39269099
Price (SGD)
126.00
PK
Clean Claims:
RNase/DNase, DNA, pyrogen and endotoxin free
Autoclavable:
Not Autoclavable
Sterility:
Non-sterile
Price (SGD)
126.00
PK
Thermo Scientific™ QSP™ Screw Cap Micro Tubes robust design and material selection are well suited for work with biologics and offer rich features, including ultra-clear externally threaded tubes and UV protective amber tubes.
Packaged in bulk bags and non-sterile clean claims are; RNase/DNase, DNA, pyrogen and endotoxin free.
  • Screw Cap micro tube volume range of 0.5, 1.5 and 2.0ml and are ideal for sample preparation, storage and transportation
  • Conical bottom or self standing offered with knurls in ultra clear natural or UV protective amber color
  • Tether caps and standard non tether caps are available in a rainbow of color options
Specifications
AutoclavableNot Autoclavable
ColorClear
MaterialPolypropylene
Clean ClaimsRNase/DNase, DNA, pyrogen and endotoxin free