Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Actual product may vary
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
Maxisafe 2030i Biological Safety Cabinets to EN12469 & DIN12980
For CMR (carcinogenic, mutagenic or toxic to reproduction) compounds. New autonomous safety features for ultimate protection and graphic user interface for ease of operation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H) ExteriorOutletsDescription
51032717870 x 1600 x 1536 mm4 Rear WallMaxisafe 2030i, 1.5 m, with 200 mm Sash Opening, with Cross Beam UV-C in Side Walls.
51032716870 x 1300 x 1536 mm2 Rear WallMaxisafe 2030i, 1.2 m, with 200 mm Sash Opening, with Cross Beam UV-C in Side Walls.
Catalog number 51032717
Price (SGD)
39,193.00
EA
Add to cart
Dimensions (L x W x H) Exterior:
870 x 1600 x 1536 mm
Outlets:
4 Rear Wall
Description:
Maxisafe 2030i, 1.5 m, with 200 mm Sash Opening, with Cross Beam UV-C in Side Walls.
Price (SGD)
39,193.00
EA
Add to cart
The Thermo Scientific™ Maxisafe 2030i Cytotoxic Safety Cabinet incorporates SmartFlow Plus, DAVe Plus and Smart UV technologies for ultimate protection and graphical user interface for ease of operation. Each unit has inbuilt Wifi capability and is ready to be connected. Our prefilter system includes a new improved and easy sealing solution. With their triple-filter design, the Maxisafe biosafey cabinet provides 100,000x better filtration efficiency than regular Class II biological safety cabinets.
Features:
  • Triple-filter design - appropriate for work with cytotoxic materials
  • Touchscreen and connectivity (remote app functionality is available in select countries) – Built-in Wifi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more.
  • Motorized front window and SmartClean Plus design - Entire front sash is motorized to either open automatically to the right aperture or to completely lift up, allowing for easy access to interior of BSC. Routine maintenance, cleaning and set up tasks are simplified.
  • Dual DC Motors and LED lights - for energy efficient operation
  • 316L Stainless Steel Interior
  • Glass side walls - for a brighter work area
  • SmartFlow Plus and DAVe Plus technologies - for enhanced protection
  • SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs
  • Options: UV crossbeam lighting, ergonomic gel-filled armrests, customizable worktops
  • Previous models: Herasafe KSP
Specifications
DescriptionMaxisafe 2030i, 1.5 m, with 200 mm Sash Opening, with Cross Beam UV-C in Side Walls.
Certifications/ComplianceEN 12469, DIN 12980
Chamber MaterialStainless Steel (316)
Product TypeBiosafety Cabinet
ModelMaxisafe 2030i
Outlets4 Rear Wall
Sash Opening (Metric)200 mm
Width (English) Exterior62.9 in.
Display7 in. GUI
Filter Efficiency0.99995
Filter TypeH14 HEPA
Green FeaturesEnergy efficient
Width (Metric) Exterior1600 mm
Energy Consumption (Reduced Flow Mode)80 W/h
LightingLED
Ports2 x Each Side plus Smart Ports
Sash Opening (English)7.8 in.
Dimensions (L x W x H) Exterior870 x 1600 x 1536 mm
Frequency50/60 Hz
Depth (Metric) Exterior870 mm
Net Weight (Metric)280 Kg
Shipping Weight (Metric)310 Kg
Voltage230 V
Energy Consumption (Operating Set Point)390 W/h
Unit SizeEA