Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Actual product may vary
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Heratherm™ General Protocol Ovens
Perform routine, day-to-day work in these general protocol ovens, which provide a safe, ideal heating and drying solution for research, clinical or industrial needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)TypeElectrical RequirementsDimensions (L x W x H)
51029342774 LGravity Convection, 3-Phase Model400 V 50/60 Hz30.3 x 49.7 x 60.8 in. (77 x 126.1 x 154.5 cm)
51028113105 LGravity Convection208/240 V 60 Hz22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
51028114176 LGravity Convection208/240 V 60 Hz29.1 x 25.2 x 36.2 in. (73.8 x 64 x 92 cm)
51028122104 LMechanical Convection Oven208/240 V 60 Hz22.2 x 25.2 x 32.3 in. (56.5 x 64 x 82 cm)
Catalog number 51029342
Price (SGD)
13,621.00
EA
Add to cart
Capacity (Metric):
774 L
Type:
Gravity Convection, 3-Phase Model
Electrical Requirements:
400 V 50/60 Hz
Dimensions (L x W x H):
30.3 x 49.7 x 60.8 in. (77 x 126.1 x 154.5 cm)
Price (SGD)
13,621.00
EA
Add to cart
Perform routine applications simply and safely with Thermo Scientific™ Heratherm™ General Protocol Ovens. These ovens provide an ideal heating and drying solution for research, clinical or industrial needs.
Features:
  • Operating temperatures from 50°C to 250°C
  • Automatic over-temperature alarm system
  • Low overall energy consumption
  • Built-in timer
  • Corrosion-resistant stainless steel chambers with rounded corners
  • Large, easy-to-read vacuum fluorescent display
  • Easy-to-use, microprocessor-controlled touch button operation
  • Doors can be opened at over 180°C
Ordering Information:
Availability may vary by country
Warranty: 2-year warranty
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Controller TypeMicroprocessor control with vacuum fluorescent display
Capacity (Metric)774 L
Temperature (Metric)50°C to 250°C
TypeGravity Convection, 3-Phase Model
No. of Shelves2 supplied/39 max.
Temperature Stability±0.5°C
TimerON / OFF
Dimensions (D x W x H) Interior23.2 x 39.5 x 51.5 in. (59 x 100.4 x 130.7 cm)
Dimensions (L x W x H)30.3 x 49.7 x 60.8 in. (77 x 126.1 x 154.5 cm)
Length (English)30.3 in.
Length (Metric)77 cm
Width (English)49.7 in.
Width (English) Interior39.5 in.
Width (Metric)126.1 cm
Width (Metric) Interior100.4 cm
Height (English)60.8 in.
Height (English) Interior51.5 in.
Height (Metric)154.5 cm
Height (Metric) Interior130.7 cm
Depth (English) Interior23.2 in.
Depth (Metric) Interior59 cm
Electrical Requirements400 V 50/60 Hz
Voltage400 V
Wattage6350 w
Amperage13 A