CryoBoxes™
Actual product may vary
CryoBoxes™
CryoBoxes™
CryoBoxes™
CryoBoxes™
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHoldsMaterial
5026-1010Blue100 (1 to 1.5 mL Vials)Polycarbonate
330821White64 (1.5 mL Microcentrifuge Tubes)Polycarbonate
FIS12-565-227White64 (1.5 mL Microcentrifuge Tubes)Polycarbonate
FIS12-567-04White100 (3.6 mL Vials)Cardboard
341483White100 (3.6 mL Vials)Polycarbonate
FIS12-565-234White100 (3.6 mL Vials)Polycarbonate
NNU341483White100 (3.6 mL Vials)Polycarbonate
369639White81 (1 to 1.8 mL Vials)Cardboard
FIS12-567-02White81 (3.6 mL Vials)Cardboard
374187Gray100 (1 to 1.8 mL Vials)Polycarbonate
FIS12-565-182Brown81 (1 to 1.8 mL Vials)Cardboard
5025-0505White25 (1.2 and 2 mL Vials)Polycarbonate
FIS15-350-107White25 (1.2 and 2 mL Vials)Polycarbonate
FKS10739331White25 (1.2 and 2 mL Vials)Polycarbonate
5026-0909White81 (1.2 and 2 mL Vials)Polycarbonate
FIS15-350-107BWhite81 (1.2 and 2 mL Vials)Polycarbonate
FIS03-337-7AABlue100 (1 to 1.5 mL Vials)Polycarbonate
FIS03-337-7JWhite81 (5 mL Vials)Polycarbonate
867013-0242Green81 (1 to 2 mL Vials)Polycarbonate
867013-0243Blue81 (1 to 2 mL Vials)Polycarbonate
867013-0245Assorted81 (1 to 2 mL Vials)Polycarbonate
FIS14-175-65Assorted81 (1 to 2 mL Vials)Polycarbonate
FKS10151042Assorted81 (1 to 2 mL Vials)Polycarbonate
Catalog number 5026-1010
HSN 39231090
Price (SGD)
226.00
CS
Color:
Blue
Holds:
100 (1 to 1.5 mL Vials)
Material:
Polycarbonate
Price (SGD)
226.00
CS
Meet the needs of any budget with plastic or cardboard CryoBoxes™ designed to store cryogenic vials and tubes. Choose from a variety of durable plastic Thermo Scientific™ CryoBox™ styles with keyed lids and printed location grids, or select economical cardboard/chipboard CryoBoxes™ for short-term storage. The plastic boxes are optimized to fit our own brand of Nalgene™ and Nunc™ vials and microcentrifuge tubes best.
  • Durable, all-plastic CryoBoxes™ for short- or long-term storage
  • Keyed lids on plastic CryoBoxes™ prevent misalignment
  • Plastic boxes are autoclavable for decontamination and usable at temperature range from -196°C to +121°C
  • Printed grid on plastic lids for easy inventory management
  • Economical chipboard/cardboard CryoBoxes™ for short term storage
  • Chipboard/cardboard boxes have dividers and vents in base for good drainage and air circulation
  • Available with dividers from 25 to 169 tube storage
Specifications
DescriptionPolycarbonate, 133 x 133 x 52mm
ColorBlue
MaterialPolycarbonate
ShapeSquare
TypeCryo Box
Holds100 (1 to 1.5 mL Vials)
Product LineNalgene
Quantity10/Pk., 10/Cs.
Tube Array10 x 10
Dimensions (L x W x H)133 x 133 x 52 mm
Length (English)5.25 in.
Length (Metric)133 mm
Width (English)5.25 in.
Width (Metric)133 mm
Height (English)2.06 in.
Height (Metric)52 mm
Weight (Metric)1.92 kg
Unit SizeCS
Frequently asked questions (FAQs)
Can I store the Thermo Scientific 2D barcoded storage tubes in standard 10x10 cryoboxes?
Yes, the Thermo Scientific 2D barcoded storage tubes can be stored in standard cryoboxes. However, we do not carry 2D barcode readers capable of reading a full box of 2D barcodes in this storage format.