Height Adjustable Floor Stand for Herasafe™ 2030i
Height Adjustable Floor Stand for Herasafe™ 2030i
Actual product may vary
Height Adjustable Floor Stand for Herasafe™ 2030i
Height Adjustable Floor Stand for Herasafe™ 2030i
Height Adjustable Floor Stand for Herasafe™ 2030i
For use with Herasafe 2030i Biological Safety Cabinets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLength (Metric) Exterior
50155691Adjustable Floor Stand (Height of 680-880mm), 1.8 m, charcoal grey1900 mm
50155689Adjustable Floor Stand (Height of 680-880mm), 1.2 m, charcoal grey1300 mm
Catalog number 50155691
Price (SGD)
2,812.00
EA
Add to cart
Description:
Adjustable Floor Stand (Height of 680-880mm), 1.8 m, charcoal grey
Length (Metric) Exterior:
1900 mm
Price (SGD)
2,812.00
EA
Add to cart
Height Adjustable Floor Stand in increments of 50mm.(Height range 680-880mm), charcoal grey
  • Adjustment steps of 50mm
  • Charcoal grey
  • Height - 680-880mm
Specifications
DescriptionAdjustable Floor Stand (Height of 680-880mm), 1.8 m, charcoal grey
Product TypeHeight Adjustable Floor Stand
Height (English)26.7 in. to 34.6 in.
Height (Metric)680 to 880 mm
Length (Metric) Exterior1900 mm
For Use With (Equipment)Herasafe 2030i Biological Safety Cabinet size 1.8 m
Depth (Metric)800 mm
Unit SizeEA