4-20mA Data Output
4-20mA Data Output
Actual product may vary
4-20mA Data Output
Converts digital actual values displayed for temperature, CO2 concentration, O2 concentration (optional) and relative humidity into a continuous output current of 4-20 mA each. The interface port accepts various customer-supplied external measurement devices with 4-20 mA inputs.
Have Questions?
Catalog NumberNo. of Places
501481408
Catalog number 50148140
Price (SGD)
899.00
EA
Add to cart
No. of Places:
8
Price (SGD)
899.00
EA
Add to cart
Converts digital actual values displayed for temperature, CO2 concentration, O2 concentration (optional) and relative humidity into a continuous output current of 4-20 mA each. The interface port accepts various customer-supplied external measurement devices with 4-20 mA inputs.
  • 4-20mA Data Output, for CO2 Incubators Heracell VIOS and Forma Steri-Cycle Models.
  • Qualified Service Installation.
  • The optional 4-20 mA interface converts digital actual values displayed for temperature, CO2 concentration, O2 concentration (optional) and relative humidity into a continuous output current of 4-20 mA each.
  • The interface port accepts various customer-supplied external measurement devices with 4-20 mA inputs.
  • 4-20 mA output signals are then broken out onto 8-pin terminals.
Specifications
Capacity (Metric)4 to 20 mA
CO2 Concentration RangeYes
For Use With (Equipment)CO2 Incubators Heracell VIOS and Forma Steri-Cycle Models
LightingNo Lighting
No. of Places8
Oxygen ControlOptional 1 to 21% or 5 to 90%
Relative HumidityYes
TypeData Output
Unit SizeEA