Specifications
Conductivity0.055μS/cm
Bacterial Content<0.01 CFU/mL
Feedwater ConnectorR 3/4 in.
Feed Water MonitoringYes
Feedwater SourcePretreated
Flow Rate1.5 L/min.
Product Water TypeType 1
Resistivity18.2 Mohm.cm
TOC Monitor with UV Intensity MonitoringNo
Total Organic Carbon1 to 5 ppb
TypeWater Purification System
Feedwater Pressure1 to 6 bar
IncludesUV-photo-oxidation
ModelMicroPure UV
Operating Pressure0.1 to 6 bar
Particles >0.2µM/mL<1 μM/mL
Power Consumption0.06 kW
Temperature Range (Metric)2°C to 35°C
Width (Metric)305 mm
Height (Metric)545 mm
Depth (Metric)300 mm
Net Weight (Metric)17 kg
Electrical Requirements100/240 V 50/60 Hz
Voltage100/240 V