Check Order Status
Support Stands for Thermo Scientific™ Heratherm Ovens and Incubators
Actual product may vary
Thermo Scientific
Support Stands for Thermo Scientific™ Heratherm Ovens and Incubators
Support stands with casters
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
50127743
180 L Model Ovens and Incubators
50127742
100 L Model Ovens and Incubators
2 Options
Catalog number
50127743
HSN
73269099
Price (SGD)
1,397.00
EA
Add to cart
For Use With (Equipment):
180 L Model Ovens and Incubators
Price (SGD)
1,397.00
EA
Add to cart
Support stands with casters
Height including casters: 7.36 in. (18.7cm).
Specifications
For Use With (Equipment)
180 L Model Ovens and Incubators
Type
Support Stand
Includes
Caster