Cimarec™ Mobil Direct Stirrers
Cimarec™ Mobil Direct Stirrers
Actual product may vary
Cimarec™ Mobil Direct Stirrers
Perform high-volume applications with the exceptional stirring power of these mobil direct stirrers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacityVoltagePlug TypeDimensions (L x W x H)
50119109200 L80 x 180 x 80 mm (7 x 7 x 3.4 in.)
50100547150 L230 VUK330 x 330 x 80 mm
Catalog number 50119109
Price (SGD)
4,464.00
EA
Add to cart
Capacity:
200 L
Dimensions (L x W x H):
80 x 180 x 80 mm (7 x 7 x 3.4 in.)
Price (SGD)
4,464.00
EA
Add to cart
Perform high-volume applications with the exceptional stirring power of Thermo Scientific™ Cimarec™ Mobil Direct Stirrers. The Mobil 200 and 600 require either the Telemodul 40 M or the Telemodul 80 M control unit.
Features:
  • Quick Stop function stops the drive within three seconds, minimizing after runs and risk of vessel breakage
  • Easy-to-clean, chemical-resistant, sealed stainless-steel housing
  • Stir speeds from 100rpm to 1000rpm
Mobil Direct:
  • Stir up to 150L
  • Maximum shaft torque is 30Ncm
  • Features an integrated controller with adjustable power settings
  • Operating conditions are -10°C to +40°C at 95%RH
  • IP64 protection rated; easily cleaned with running water
Mobil 200:
  • Stir up to 200L
  • Maximum shaft torque is 50Ncm
  • Operating conditions are -10°C to +56°C at 100%RH
  • IP65 protection rated; easily cleaned with running water
Mobil 600:
  • Stir up to 600L
  • Maximum shaft torque is 50Ncm
  • Operating conditions are -10°C to +56°C at 100%RH
  • IP65 protection rated; easily cleaned with running water
Requires:
The Mobil 200 and 600 require either the Telemodul 40M or 80M controller (order separately). The Mobil Direct includes a built-in controller; no external controller required.
Recommended for:
Use in pilot plant or integrated with process equipment
Warranty: 5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed100 to 1000 rpm
Top Plate MaterialStainless Steel
Capacity200 L
ShapeSquare
Wattage80 w
Dimensions (L x W x H)80 x 180 x 80 mm (7 x 7 x 3.4 in.)
Certifications/ComplianceCE
DescriptionMobil 200 w/o Controller; Capacity 200L
ModelMobil 200 without Controller
Protection ClassIP64, IP32
Temperature Range (Metric)-10°C to +40°C (Operating)
TypeStirrer
Weight (English)13.2 lb.
Weight (Metric)6 kg
Unit SizeEA