Cimarec™ Biosystem 40B Controller
Cimarec™ Biosystem 40B Controller
Actual product may vary
Cimarec™ Biosystem 40B Controller
Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.
Have Questions?
Catalog NumberVoltage
50118918230 V UK
Catalog number 50118918
Price (SGD)
2,999.00
EA
Add to cart
Voltage:
230 V UK
Price (SGD)
2,999.00
EA
Add to cart
Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.
  • High visibility display
  • Adjustable stirring and pause times with rotation sense inversion
Specifications
DescriptionBiomodul 40B Controller 230V UK
ModelBiomodul 40B Controller
TypeStirrer Controller (Dual)
Voltage230 V UK
Electrical Requirements230 V
Unit SizeEA