Teleshake Magnetic Shaking System
Teleshake Magnetic Shaking System
Actual product may vary
Teleshake Magnetic Shaking System
Teleshake Magnetic Shaking System
Teleshake Magnetic Shaking System
Conduct powerful shaking applications from 100 to 2000rpm with this remote-controlled, wear-free magnetic shaking system.
Have Questions?
Catalog NumberVoltage
50095890230 V (UK)
Catalog number 50095890
Price (SGD)
5,044.00
EA
Add to cart
Voltage:
230 V (UK)
Price (SGD)
5,044.00
EA
Add to cart
Conduct powerful shaking applications from 100 to 2000rpm with the Thermo Scientific™ Teleshake Magnetic Shaking System, a remote-controlled, wear-free magnetic shaking system for integrated shaking applications; suited for 96- and 384-well microtiter plates.
  • Easy integration in robotic systems and platform due to small footprint
  • Simultaneous control of up to 14 shakers via RS-232 interface
  • Powerful shaking movements from 100 to 2000rpm, with an amplitude of 2mm
  • Rapid switching between circular and linear agitation
  • Minimum of moved masses, therefore little transference of vibrations onto the installation area
  • Integrated threads and drilled holes for assembling on robot workbenches
Specifications
Speed Range100 to 2000 rpm
Voltage230 V (UK)
Temperature Range (Metric)5°C to 40°C
Amplitude2 mm
Product TypeShaker
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Height (English)1.85 in.
Height (Metric)47 mm
Width (English)5.74 in.
Width (Metric)146 mm
Humidity Range95% at 5° to 40°C
IncludesControl unit, 230V UK
No. of Positions1
Unit SizeEA