Cimarec™ i Test Tube Racks
Cimarec™ i Test Tube Racks
Actual product may vary
Cimarec™ i Test Tube Racks
Use Thermo Scientific™ Cimarec™ i Test Tube Racks with Telesystem Series Stirrers.
Have Questions?
Catalog NumberFor Use With (Equipment)
50087960
also known as 50087960J
25mm tubes in Telesystem Series Stirrers
Catalog number 50087960
also known as 50087960J
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
25mm tubes in Telesystem Series Stirrers
Use Thermo Scientific™ Cimarec™ i Test Tube Racks with Telesystem Series Stirrers.
  • Stainless-steel test tube racks ensure test tubes are precisely centered over stirring points
Specifications
For Use With (Equipment)25mm tubes in Telesystem Series Stirrers
TypeTest Tube Rack