Check Order Status
Filter Disc, Surfaces Plane, Ground Cylindrical, 270 mm
Filter Disc, Surfaces Plane, Ground Cylindrical, 270 mm
The faces of the VitraPOR™ biplane filter discs from series 16 are precisely ground down to tight tolerances and plane surfaces.
Have Questions?
Catalog Number
Porosity
50062701
also known as GT50062701
1
Catalog number
50062701
also known as GT50062701
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Porosity:
1
Highly resistant to most chemicals and temperatures up to 500°C
The biplane version may be required in specific installation circumstances
Specifications
Material
Borosilicate Glass (3.3)
Pore Size
1 μm
Sterility
Non-sterile
No. per Pack
1 Ea.
Porosity
1
Thickness (Metric)
18.0 mm ± 0.5 mm
Diameter (Metric)
270 mm