PTFE Platinum Restistance Temperature Probe, 9mm Diameter
Product Image
PTFE Platinum Restistance Temperature Probe, 9mm Diameter
Fisherbrand™ PTFE Platinum Restistance Temperature Probes combine the outstanding properties of PTFE with the reliability and accuracy of platinum resistance thermometry. All components are fully encapsulted in pure PTFE. The body has a stainless steel core for rigidity but can be bent.
Have Questions?
Catalog NumberDimensions (L x W)Length (Metric)
5005-0015500 x 9 mm5 m (Cable)
Catalog number 5005-0015
Price (SGD)
196.00
CS
Add to cart
Dimensions (L x W):
500 x 9 mm
Length (Metric):
5 m (Cable)
Price (SGD)
196.00
CS
Add to cart
  • Sensor - PRT100 Class A. TCR = 3850 ppm/°C.
  • Cable - 4 Wire; 2 Red, 2 White, PFA insulated with white PFA oversheath
  • Cable fused to body
  • Response - 50%, 6-14 seconds.
  • With push on ferrule
  • Supplied without plug
  • Inert and non contaminating
  • Pressure Resistant
  • Temperature Range -180°C to +280°C
Specifications
DescriptionFisherbrand™ PTFE Platinum Restistance Temperature Probe
MaterialPTFE with PRT100 Sensor
Quantity1
Sensor TypesPRT100 Class A. TCR = 3850 ppmC
Response Time50%, 6 to 14 sec.
Temperature Range (Metric)-180°C to +280°C
Dimensions (L x W)500 x 9 mm
Length (Metric)5 m (Cable)
Width (Metric)9 mm
Unit SizeCS