Diaphragm Pump HMD 8
Diaphragm Pump HMD 8
Actual product may vary
Diaphragm Pump HMD 8
Three Step Diaphragm Pump. Standard Pump for Drying (Water/ Non-Caustic Vapours). Supplied Complete with All Necessary Tubing and Fittings to Connect to the Vacuum Oven.
Have Questions?
Catalog number 50040078
Price (SGD)
25,287.00
EA
Add to cart
Price (SGD)
25,287.00
EA
Add to cart
Three Step Diaphragm Pump. Standard Pump for Drying (Water/ Non-Caustic Vapours). Supplied Complete with All Necessary Tubing and Fittings to Connect to the Vacuum Oven.
  • Condensate trap and drain cock
  • 100 % oil free, requires little maintenance
  • Powerful
  • Short pump-down times
Specifications
Max. Vacuum3.37 kW
Certifications/ComplianceCE certified
Dimensions (L x W x H)610 x 240 x 300 mm
ModelHMD 8
Weight (Metric)26 kg
Flow Rate9.6 m3/h
Pump TypeVacuum Pump
Enclosure RatingIP20
ApplicationsFor Drying (Water/ Non-Caustic Vapours)
Maximum Pressure<10 Mbar final pressure with gas ballast
For Use With (Equipment)Vacutherm 6000 Oven