Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Actual product may vary
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Plates and Modules with Covalent Binding Surfaces
Reduce the amount of reagents used in the lab by using these plates and modules with covalent binding surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorVolume (Metric) WorkingProduct TypeSurface Treatment
478042Clear100 μLSolid Module F8 (x12) in FrameCovalent Binding
436006
also known as GT436006
Clear100 μLMicroplateCovalent Binding
436023Clear100 μLMicroplateAmine
Catalog number 478042
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Clear
Volume (Metric) Working:
100 μL
Product Type:
Solid Module F8 (x12) in Frame
Surface Treatment:
Covalent Binding
Reduce the amount of reagents used in the lab with Thermo Scientific™ Plates and Modules with Covalent Binding Surfaces.
Covalent Surfaces (Thermo Scientific™ CovaLink™ and Immobilizer Amino)
  • Minimal leaching
  • Withstands vigorous washing
  • Coating with lower amounts of reagent may be possible
  • Control of orientation
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Immobilizer is a trademark of Exiqon A/S, Vedbaek, Denmark. The product is produced under license from Exiqon A/S and covered by EP 08 20483 and foreign application and patents.
Specifications
FormatStandard
Detection MethodColorimetric
Array8 x 12
Packaging30 Cases
ColorClear
Product TypeSolid Module F8 (x12) in Frame
MaterialPolystyrene
Volume (Metric) Well400 μL
No. of Wells96
Volume (Metric) Working100 μL
Surface TreatmentCovalent Binding
Total Volume (Metric) Well400 μL
Well ShapeRound, Flat Bottom (F-well)
No. per Case30
No. per Pack5
Plate BlockingUnspecified
Target MoleculePeptides
Unit SizeCS